Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6207

Sigma-Aldrich

sCD23 human

recombinant, expressed in E. coli, ≥96% (SDS-PAGE)

Synonym(s):

BLAST-2, CD23 antigen, Fc-epsilon-RII, Lymphocyte IgE receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

assay

≥96% (SDS-PAGE)

form

lyophilized powder

mol wt

19.2 kDa

packaging

pkg of 10 μg
pkg of 50 μg

impurities

<1 EU/μg endotoxin (LAL test)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CD23(9606)

General description

CD23, the low affinity receptor for IgE, belongs to the C-type lectin structural family and plays a role in the regulation of IgE synthesis and IgE mediated activities. It is found both as a transmembrane receptor protein and in a soluble form, which is generated by proteolytic cleavage of membrane bound CD23. The predominant soluble form of CD23 (sCD23) consists of 172 amino acids corresponding to the extracellular domain of the full length precursor. sCD23, in addition to binding IgE, also exerts a number of IgE independent activities, such as promoting the activation and differentiation of B-cells and stimulating the release of pro-inflammatory cytokines from monocytes. Recombinant human sCD23 is a 19.2 kDa non-glycosylated protein containing 172 amino-acid residues.

Physical form

Sterile filtered through a 0.2 micron filter. Lyophilized from 0.5X PBS, pH 8.0.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service