Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6193

Sigma-Aldrich

Hemocyanin-Keyhole Limpet subunits

Synonym(s):

Megathura crenulata hemocyanin, keyhole limpett hemocyanin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

form

lyophilized

mol wt

350 and 390 kDa (Two main characteristic bands co-migrating with ferritin)

packaging

pkg of 1 mg

shipped in

wet ice

storage temp.

2-8°C

General description

Hemocyanins are proteins that use copper binding sites to bind and transport oxygen in a variety of arthropods and mollusks. Hemocyanin is isolated from the hemolymph of the animals.[1] It is one of the strongest antigens known. Hemocyanin has been in use as an immunological reagent for many years. It is used as a carrier protein for antibody production against antigens. Recent advances in immunology and the role immune system plays in diseases have opened a whole new era of product development activities aimed at developing novel therapeutics which is aimed at teaching the body′s immune system to fight diseases like cancer, AIDS, etc. The approach involves the use of highly immunogenic molecule like the hemocyanin for non-specific immunostimulation (NSI) or active specific immunostimulation (ASI) using conjugate vaccines, wherein the tumor (disease) specific antigens are covalently bound to carrier protein like KLH and the product used in human clinical studies. Such products are termed “vaccines”. The KLH subunits powder has major advantages associated with it, in terms of flexibility of use and the choice of buffer in early developmental studies. These subunits are highly pure and have low endotoxin content.

Physical form

Freeze Dried.

Reconstitution

Resuspend in sterile water to a 1 mg/mL concentration.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hemocyanins and invertebrate evolution.
van Holde KE
The Journal of Biological Chemistry, 276(19), 15563-15566 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service