Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6165

Sigma-Aldrich

M-CSF human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

CSF-1, Lanimostim, MGC31930, Macrophage colony stimulating factor 1

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in HEK 293 cells

assay

≥95% (SDS-PAGE)

form

lyophilized

mol wt

(35 and 40 kDa, homodimer, glycosylated)

packaging

pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CSF1(1435)

General description

Granulocyte/macrophage colony-stimulating factors (MCSFs) are cytokines that act in hematopoiesis by controlling the production, differentiation, and function of 2 related white cell populations of the blood, the granulocytes and the monocytes-macrophages. MCSF induces cells of the monocyte/macrophage lineage. MCSF plays a role in immunological defenses, bone metabolism, lipoproteins clearance, fertility and pregnancy.

Biochem/physiol Actions

The gene encoding macrophage colony-stimulating factor (MCSF) is located on human chromosome 1p13.3. It is a glycosylated polypeptide which exists as a homodimer.[1][2]

Physical form

Lyophilized from a PBS solution.

Preparation Note

Centrifuge the vial prior to opening.

Reconstitution

Reconstitute in sterile PBS containing 0.1% endotoxin-free, recombinant human serum albumin.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genome-wide association study identifies variants at CSF1, OPTN and TNFRSF11A as genetic risk factors for Paget's disease of bone.
Albagha OM
Nature Genetics, 42(6), 520-524 (2010)
Synthesis of membrane-bound colony-stimulating factor 1 (CSF-1) and downmodulation of CSF-1 receptors in NIH 3T3 cells transformed by cotransfection of the human CSF-1 and c-fms (CSF-1 receptor) genes.
Rettenmier CW
Molecular and Cellular Biology, 7(7), 2378-2387 (1987)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service