SRP6162
FGF-8b human
recombinant, expressed in human cells, ≥95% (SDS-PAGE)
Synonym(s):
AIGF, FGF-17a, Fibroblast growth factor 17a, Fibroblast growth factor 8b, HBGF-8
About This Item
Recommended Products
biological source
human
recombinant
expressed in human cells
assay
≥95% (SDS-PAGE)
form
lyophilized
mol wt
30-45 kDa (monomer, glycosylated)
packaging
pkg of 10 μg
technique(s)
cell culture | mammalian: suitable
NCBI accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... FGF8(2253)
General description
Biochem/physiol Actions
Physical form
Preparation Note
Reconstitution
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service