Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6150

Sigma-Aldrich

UBE2E2 human

≥95% (SDS-PAGE)

Synonym(s):

MGC40331, RIG-B, UBE2L6, UbcH8, Ubiquitin-protein ligase L6, Ubiquitin/ISG15-conjugating enzyme E2 L6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

assay

≥95% (SDS-PAGE)

form

liquid

mol wt

22.255 kDa

packaging

pkg of 3 nmol

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... UBE2E2(7325)

General description

Ubiquitin-conjugating enzyme E2 (UBE2E2) is a protein that in humans is encoded by the UBE2E2 gene. Amino acid sequence within the UBC domain of UBE2E2 shares over 90% identity with human UbcH6, mouse UbcM2, and Drosophila UbcD2, whereas the N-terminal region shows little amino acid sequence similarity with known proteins. The UBE2E2 protein forms a thioester bond with ubiquitin in an E1-dependent manner and mediates the transfer of ubiquitin from a ubiquitin-activating enzyme (E1) to a substrate protein or E3 ligase.

Physical form

75 μL of a 40 μM solution (3 nmoles) in 20 mM Tris, pH 7.4, 150 mM NaCl, 10 mM DTT and 10% glycerol.

Preparation Note

Centrifuge the vial prior to opening.

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Min Deng et al.
Science advances, 6(37) (2020-09-13)
DNA double-strand breaks (DSBs) are highly toxic lesions that can drive genetic instability. These lesions also contribute to the efficacy of radiotherapy and many cancer chemotherapeutics. DNA repair efficiency is regulated by both intracellular and extracellular chemical signals. However, it

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service