Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6120

Sigma-Aldrich

Fumarase human

recombinant, expressed in E. coli, ≥95% (SDS-PAGE)

Synonym(s):

FH, Fumarate hydratase, LRCC, MCL, MCUL1

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

assay

≥95% (SDS-PAGE)

form

liquid

potency

>1.0 units/mg

mol wt

50.2 kDa (467 aa, 44-510 aa)

packaging

pkg of 100 μg

impurities

<1.0 EU/μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... FH(2271)

General description

Fumarase (Fumarate hydratase) is an enzyme that catalyzes the reversible hydration/dehydration of fumarate to S-malate and is involved in the tricarboxylic acid (TCA), or Krebs cycle. This enzyme exists in both a cytosolic form and an N-terminal extended mitochondrial form. The N-terminal extended form is targeted to the mitochondrion, where the removal of the extension is the same form as in the cytoplasm. Fumarase deficiency can lead to progressive encephalopathy, cerebral atrophy and developmental delay and this enzyme also is thought to act as a tumor suppressor.

Physical form

1 mg/mL solution in 20 mM Tris-HCl buffer (pH 8.0).

Preparation Note

Centrifuge the vial prior to opening.

Other Notes

MASQNSFRIE YDTFGELKVP NDKYYGAQTV RSTMNFKIGG VTERMPTPVI KAFGILKRAA AEVNQDYGLD PKIANAIMKA ADEVAEGKLN DHFPLVVWQT GSGTQTNMNV NEVISNRAIE MLGGELGSKI PVHPNDHVNK SQSSNDTFPT AMHIAAAIEV HEVLLPGLQK LHDALDAKSK EFAQIIKIGR THTQDAVPLT LGQEFSGYVQ QVKYAMTRIK AAMPRIYELA AGGTAVGTGL NTRIGFAEKV AAKVAALTGL PFVTAPNKFE ALAAHDALVE LSGAMNTTAC SLMKIANDIR FLGSGPRSGL GELILPENEP GSSIMPGKVN PTQCEAMTMV AAQVMGNHVA VTVGGSNGHF ELNVFKPMMI KNVLHSARLL GDASVSFTEN CVVGIQANTE RINKLMNESL MLVTALNPHI GYDKAAKIAK TAHKNGSTLK ETAIELGYLT AEQFDEWVKP KDMLGPK

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Izanara C Pritsch et al.
BMC molecular and cell biology, 21(1), 90-90 (2020-12-09)
The zoonotic worm parasite Fasciola hepatica secretes an abundance of cathepsin L peptidases that are associated with virulence, invasiveness, feeding and migration. The peptidases are produced as inactive zymogens that activate at low pH by autocatalytic removal of their N-terminal
Carolina De Marco Verissimo et al.
PLoS neglected tropical diseases, 14(8), e0008510-e0008510 (2020-08-08)
Serine protease inhibitors (serpins) regulate proteolytic events within diverse biological processes, including digestion, coagulation, inflammation and immune responses. The presence of serpins in Fasciola hepatica excretory-secretory products indicates that the parasite exploits these to regulate proteases encountered during its development
Mark W Robinson et al.
PLoS pathogens, 7(5), e1002042-e1002042 (2011-05-19)
Over the last decade a significant number of studies have highlighted the central role of host antimicrobial (or defence) peptides in modulating the response of innate immune cells to pathogen-associated ligands. In humans, the most widely studied antimicrobial peptide is
I L Pacheco et al.
Veterinary parasitology, 238, 61-65 (2017-04-08)
The expression of IFNγ and IL4 was quantified using q-PCR in the liver and hepatic lymph nodes (HLN) of sheep during early stages of infection with Fasciola hepatica (1, 3, 9 and 18days post-infection, dpi). A group of animals (Group
Isabel L Pacheco et al.
Veterinary research, 49(1), 56-56 (2018-07-05)
The expression of T regulatory cells (Foxp3), regulatory (interleukin [IL]-10 and transforming growth factor beta [TGF-β]) and proinflammatory (tumor necrosis factor alpha [TNF-α] and interleukin [IL]-1β) cytokines was quantified using real time polymerase chain reaction (qRT-PCR) in the liver of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service