Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6109

Sigma-Aldrich

α-Enolase human

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Synonym(s):

2-phospho-D-glycerate hydro-lyase, Alpha-enolase, ENO1, Enolase 1, Non-neural enolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

assay

≥90% (SDS-PAGE)

form

liquid

mol wt

47.1 kDa (434 aa, 1-434 aa)

packaging

pkg of 100 μg

technique(s)

activity assay: suitable

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... ENO1(2023)

General description

α-Enolase is encoded by the gene mapped to human chromosome 1p36.3-p36.2.[1]
α-Enolase, also known as Enolase 1, is one of three enolase isoenzymes and a glycolytic enzyme expressed in most tissues. It is predominantly expressed in the early stages of embryonic development. The enzyme consists of two subunits of 433 amino-acids each. Each subunit has a molar mass of 47kDa. The dimer is stabilized in the presence of magnesium.[2]

Application

α-Enolase human has been used in in vitro kinase activity assay.[3]

Biochem/physiol Actions

α-Enolase catalyzes the dehydration of 2-phosphoglycerate to phosphoenolpyruvate. This protein plays a key role in anaerobic metabolism under hypoxic conditions and may act as a cell surface plasminogen receptor during tissue invasion. Abnormal expression of α-enolase is associated with tumor progression in some cases of breast and lung cancer. It also has been identified as an auto antigen associated with Hashimoto′s encephalopathy and severe asthma.[2][4]

Physical form

1 mg/mL solution in 20 mM Tris-HCl (pH 7.5) containing 10% glycerol and 1 mM MgSO4.

Preparation Note

Centrifuge the vial prior to opening.

Other Notes

MSILKIHARE IFDSRGNPTV EVDLFTSKGL FRAAVPSGAS TGIYEALELR DNDKTRYMGK GVSKAVEHIN KTIAPALVSK KLNVTEQEKI DKLMIEMDGT ENKSKFGANA ILGVSLAVCK AGAVEKGVPL YRHIADLAGN SEVILPVPAF NVINGGSHAG NKLAMQEFMI LPVGAANFRE AMRIGAEVYH NLKNVIKEKY GKDATNVGDE GGFAPNILEN KEGLELLKTA IGKAGYTDKV VIGMDVAASE FFRSGKYDLD FKSPDDPSRY ISPDQLADLY KSFIKDYPVV SIEDPFDQDD WGAWQKFTAS AGIQVVGDDL TVTNPKRIAK AVNEKSCNCL LLKVNQIGSV TESLQACKLA QANGWGVMVS HRSGETEDTF IADLVVGLCT GQIKTGAPCR SERLAKYNQL LRIEEELGSK AKFAGRNFRN PLAK

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phosphoproteomic analysis of interacting tumor and endothelial cells identifies regulatory mechanisms of transendothelial migration.
Locard-Paulet M, et al.
Science Signaling, 9(414), ra15-ra15 (2016)
Alpha-enolase: a target of antibodies in infectious and autoimmune diseases.
Terrier B
Autoimmunity Reviews, 6, 176-182 (2007)
Hye-Ah Lee et al.
Yonsei medical journal, 49(6), 923-930 (2008-12-25)
A possible involvement of autoimmune mechanism in the pathogenesis of bronchial asthma has been proposed. Recently, alpha-enolase protein was identified as a major autoantigen recognized by circulating IgG autoantibodies in patients with severe asthma. To evaluate a possible pathogenetic significance
a-Enolase: a promising therapeutic and diagnostic tumor target.
Capello M
FEBS Journal, 278, 1064-1074 (2011)
Effects of pulsed electromagnetic fields on human osteoblastlike cells (MG-63): a pilot study.
Sollazzo V, et al.
Clinical Orthopaedics and Related Research, 468(8), 2260-2277 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service