SRP6096
FKBPL human
recombinant, expressed in E. coli, ≥90% (SDS-PAGE)
Synonym(s):
DIR1, FK506 binding protein like, NG7, WISP39
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352200
NACRES:
NA.32
biological source
human
recombinant
expressed in E. coli
assay
≥90% (SDS-PAGE)
form
liquid
mol wt
39.2 kDa
packaging
pkg of 10 μg
NCBI accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... FKBPL(63943)
General description
FKBPL is related to the immunophilin protein family, which take part in immunoregulation and regular cellular processes involving protein folding and trafficking. FKBPL has an important function in the induced radio resistance and participates in the control of the cell cycle. FKBPL has a role in cellular response to stress. FKBPL interacts with Hsp90, glucocorticoid receptor and dynamitin and is involved in signaling, like other FKBPs.
Physical form
0.25 mg/mL solution in 20 mM Tris-HCl buffer (pH 8.0) containing 2 mM DTT, 20% glycerol, 1 mM EDTA and 100 mM NaCl.
Preparation Note
Centrifuge the vial prior to opening.
Other Notes
METPPVNTIG EKDTSQPQQE WEKNLRENLD SVIQIRQQPR DPPTETLELE VSPDPASQIL EHTQGAEKLV AELEGDSHKS HGSTSQMPEA LQASDLWYCP DGSFVKKIVI RGHGLDKPKL GSCCRVLALG FPFGSGPPEG WTELTMGVGP WREETWGELI EKCLESMCQG EEAELQLPGH SGPPVRLTLA SFTQGRDSWE LETSEKEALA REERARGTEL FRAGNPEGAA RCYGRALRLL LTLPPPGPPE RTVLHANLAA CQLLLGQPQL AAQSCDRVLE REPGHLKALY RRGVAQAALG NLEKATADLK KVLAIDPKNR AAQEELGKVV IQGKNQDAGL AQGLRKMFGL EHHHHHH
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service