Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6076

Sigma-Aldrich

PRDX 6 human

recombinant, expressed in E. coli, ≥95% (SDS-PAGE)

Synonym(s):

1-Cys, AOP2, NSGPx, Peroxiredoxin 6, aiPLA2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

assay

≥95% (SDS-PAGE)

form

liquid

mol wt

27.1 kDa

packaging

pkg of 100 μg

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PRDX6(9588)

General description

Peroxiredoxin 6 (Prdx6) is a nonseleno peroxidase enzyme, encoded by the gene mapped to human chromosome 1q25.1, a region associated with schizophrenia.[1][2] Prdx6 belongs to thiol-specific antioxidant protein family.[1] Prdx6 is highly expressed in lung, alveolar epithelial type II cells, bronchiolar Clara cells, and alveolar macrophages.[3] Unlike other members of the family, Prdx6 is characterized with a single conserved cysteine residue and has been called as 1-Cys Prdx. Prdx6 is a ~25,100Da protein containing 224 amino acids.[2]

Biochem/physiol Actions

Peroxiredoxin 6 (Prdx6) is a bifunctional enzyme, carrying out both phospholipase A2 (PLA2) activity and glutathione (GSH) peroxidase function. The encoded protein aids in redox regulation of the cell and is implicated in reduction of H2O2 and short chain organic, fatty acid, and phospholipid hydroperoxides. It might also have a crucial role in the regulation of phospholipid turnover and in antioxidant defense.[1][2] Prdx6 is highly expressed at leading edges of wounds and aids in wound healing. Deficiency of the enzyme activity is observed in cataracts of the lens. Overexpression of Prdx6 leads to several degenerative CNS (central nervous system) disorders such as Alzheimer′s, Parkinson′s, Pick′s, and Crutzfeld-Jakob diseases and schizophrenia. In addition, elevated expression of the enzyme is also observed in various neoplastic diseases including malignant mesothelioma, bronchogenic squamous cell carcinoma and cancer of the gingivo-buccal area, bladder and breast.[2]

Physical form

1 mg/mL solution in 20 mM Tris-HCl buffer (pH 8.0) containing 20% glycerol.

Preparation Note

Centrifuge the vial prior to opening.

Other Notes

MGSSHHHHHH SSGLVPRGSH MPGGLLLGDV APNFEANTTV GRIRFHDFLG DSWGILFSHP RDFTPVCTTE LGRAAKLAPE FAKRNVKLIA LSIDSVEDHL AWSKDINAYN CEEPTEKLPF PIIDDRNREL AILLGMLDPA EKDEKGMPVT ARVVFVFGPD KKLKLSILYP ATTGRNFDEI LRVVISLQLT AEKRVATPVD WKDGDSVMVL PTIPEEEAKK LFPKGVFTKE LPSGKKYLRY TPQP

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peroxiredoxin 6 as an antioxidant enzyme: protection of lung alveolar epithelial type II cells from H2O2-induced oxidative stress.
Wang Y
Journal of Cellular Biochemistry, 104(4), 1274-1285 (2008)
Peroxiredoxin 6: a bifunctional enzyme with glutathione peroxidase and phospholipase A2 activities.
Fisher AB
Antioxidants & Redox Signaling, 15(3), 831-844 (2011)
Proteome analysis of schizophrenia patients Wernicke's area reveals an energy metabolism dysregulation.
Martins-de-Souza D
BMC Psychiatry, 9:17 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service