Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6075

Sigma-Aldrich

PRDX 5 human

recombinant, expressed in E. coli, ≥95% (SDS-PAGE)

Synonym(s):

ACR1, AOEB116, B116, PLP, Peroxiredoxin 5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

assay

≥95% (SDS-PAGE)

form

liquid

mol wt

17 kDa

packaging

pkg of 100 μg

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PRDX5(25824)

General description

Peroxiredoxin-5, mitochondrial in humans is encoded by the PRDX5 gene. This gene encodes a member of the peroxiredoxin family of antioxidant enzymes, which reduce hydrogen peroxide and alkyl hydroperoxides. The encoded protein may play an antioxidant protective role in different tissues under normal conditions and during inflammatory processes. It interacts with peroxisome receptor 1. In human cells, it has been shown that PRDX5 can be addressed to mitochondria, peroxisomes, the cytosol, and the nucleus.

Physical form

1 mg/mL solution in 20 mM HEPES buffer (pH 7.4).

Preparation Note

Centrifuge the vial prior to opening.

Other Notes

MAPIKVGDAI PAVEVFEGEP GNKVNLAELF KGKKGVLFGV PGAFTPGCSK THLPGFVEQA EALKAKGVQV VACLSVNDAF VTGEWGRAHK AEGKVRLLAD PTGAFGKETD LLLDDSLVSI FGNRRLKRFS MVVQDGIVKA LNVEPDGTGL TCSLAPNIIS QL.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service