Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6069

Sigma-Aldrich

Cyclophilin E human

recombinant, expressed in E. coli, ≥95% (SDS-PAGE)

Synonym(s):

CYP-33, PPIE, PPIase E, Peptidyl prolyl isomerase E, Rotamase E

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

assay

≥95% (SDS-PAGE)

form

liquid

mol wt

37.5 kDa

packaging

pkg of 100 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PPIE(10450)

General description

Cyclophilins possess the activity of peptidyl-prolyl cis-trans isomerase (PPIase) and are involved in cellular protein folding and protein interactions. PPIE contains two RNA binding domains at the N-terminal region and a PPIase domain at the C-terminal region. The cellular and physiological roles of PPIE are still not known.

Physical form

1 mg/mL solution in 20 mM Tris-HCl buffer (pH 8.0)

Preparation Note

Centrifuge the vial prior to opening.

Other Notes

MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSMATT KRVLYVGGLA EEVDDKVLHA AFIPFGDITD IQIPLDYETE KHRGFAFVEF ELAEDAAAAI DNMNESELFG RTIRVNLAKP MRIKEGSSRP VWSDDDWLKK FSGKTLEENK EEEGSEPPKA ETQEGEPIAK KARSNPQVYM DIKIGNKPAG RIQMLLRSDV VPMTAENFRC LCTHEKGFGF KGSSFHRIIP QFMCQGGDFT NHNGTGGKSI YGKKFDDENF ILKHTGPGLL SMANSGPNTN GSQFFLTCDK TDWLDGKHVV FGEVTEGLDV LRQIEAQGSK DGKPKQKVII ADCGEYV

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service