Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6059

Sigma-Aldrich

Progranulin from mouse

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)

Synonym(s):

PC cell-derived growth factor, PEPI, Proepithelin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

mouse

recombinant

expressed in HEK 293 cells

assay

≥95% (SDS-PAGE)

form

liquid

mol wt

~70.0 kDa

packaging

pkg of 10 μg

manufacturer/tradename

Sigma-Aldrich

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... GRN(10090)

General description

tumor suppression and are expressed in a cell-specific manner. Plasminogen activator inhibitor 1, also known as PAI-1, Endothelial plasminogen activator inhibitor, SERPINE1 and

Physical form

0.2 μm-filtered solution in PBS

Preparation Note

Centrifuge the vial prior to opening.

Other Notes

Signal peptide and mouse Progranulin (aa 1-589) fused at the C-terminus to a FLAG®-tag.

Legal Information

FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lorenz Thurner et al.
Digestive diseases and sciences, 59(8), 1733-1742 (2014-03-05)
Recently, we identified neutralizing autoantibodies against progranulin (PGRN) in a wide spectrum of rheumatic diseases including cases with enteropathic spondylarthritis. PGRN is a secreted protein with strong anti-inflammatory effects, believed to be mediated by the direct inhibition of TNF receptors
Masato Kanazawa et al.
Brain : a journal of neurology, 138(Pt 7), 1932-1948 (2015-04-04)
In the central nervous system, progranulin, a glycoprotein growth factor, plays a crucial role in maintaining physiological functions, and progranulin gene mutations cause TAR DNA-binding protein-43-positive frontotemporal lobar degeneration. Although several studies have reported that progranulin plays a protective role
Michael J Smout et al.
PLoS pathogens, 11(10), e1005209-e1005209 (2015-10-21)
Infection with the human liver fluke Opisthorchis viverrini induces cancer of the bile ducts, cholangiocarcinoma (CCA). Injury from feeding activities of this parasite within the human biliary tree causes extensive lesions, wounds that undergo protracted cycles of healing, and re-injury

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service