Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6055

Sigma-Aldrich

Granzyme A human

recombinant, expressed in mammalian cells, ≥95% (SDS-PAGE), ≥95% (HPLC)

Synonym(s):

CTL tryptase, Cytotoxic T-lymphocyte proteinase 1, Fragmentin-1, Granzyme-1, Hanukkah factor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in mammalian cells

assay

≥95% (HPLC)
≥95% (SDS-PAGE)

form

lyophilized

mol wt

26.9 kDa

packaging

pkg of 10 μg

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... GZMA(3001)

General description

Granzyme A is a member of the granzyme family. Granzyme A is the most abundant serine protease in cytotoxic T lymphocytes (CTL) and natural killer (NK) cells. Granzyme A has a specific function in CTL and NK cells. It induces caspase-independent cell death when introduced into target cells by perforin. Human granzyme A is synthesized as a precursor (262 residues) with a signal peptide (residues 1-26), a propeptide (residues 27-28) and a mature chain (residues 29-262).

Physical form

Lyophilized from a 0.2 μm filtered solution of 20 mM MES and150 mM NaCl, pH 5.5.

Preparation Note

Centrifuge the vial prior to opening.

Reconstitution

Dissolve in 1x PBS (It is not recommended to reconstitute to a final concentration less than 100 μg/mL.). This can further be diluted to other aqueous buffers.

Other Notes

Recombinant Human Granzyme A is produced with mammalian cells expression system. Target protein is expressed with sequence (Glu27-Val262) of Human granzyme a (Uniprot #P12544) fused with a polyhistidine tag at the C-terminus.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laura Soriano-Romaní et al.
Experimental eye research, 134, 1-14 (2015-03-11)
Recently, thrombospondin-1 (TSP-1) has been reported to be critical for maintaining a healthy ocular surface. The purpose of the study was to characterize the expression of TSP-1 and of its receptors CD36 and CD47 in corneal and conjunctival epithelial cells
Sung Min Son et al.
Neurobiology of aging, 36(12), 3214-3227 (2015-10-11)
Alzheimer's disease (AD) is characterized by impaired cognitive function and memory loss, which are often the result of synaptic pathology. Thrombospondin (TSP) is an astrocyte-secreted protein, well known for its function as a modulator of synaptogenesis and neurogenesis. Here, we
Sehwon Koh et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 35(47), 15649-15665 (2015-11-27)
Cell therapy demonstrates great potential for the treatment of neurological disorders. Human umbilical tissue-derived cells (hUTCs) were previously shown to have protective and regenerative effects in animal models of stroke and retinal degeneration, but the underlying therapeutic mechanisms are unknown.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service