SRP6042
IL-17E murine
recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC)
Synonym(s):
IL17E, IL25, Interleukin-17E, Interleukin-25
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
biological source
mouse
recombinant
expressed in E. coli
assay
≥95% (HPLC)
≥95% (SDS-PAGE)
form
lyophilized
mol wt
34.9 kDa
packaging
pkg of 25 μg
impurities
Endotoxin, tested
NCBI accession no.
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
Gene Information
mouse ... IL-17E(140806)
General description
Mouse IL-17E (IL-25), is a proinflammatory cytokine member of a six-species family of proteins (IL-17A-17F). Mouse IL-17E protein is a homodimer consisting of two 153 amino acids peptides. Recombinant mouse IL-17E produced in E.coli is a non-glycosylated disulfide-joined homodimer having a molecular mass of 34.9 kDa.
Physical form
Lyophilized with no additives
Reconstitution
Centrifuge the vial prior to opening. When reconstituting the product, gently pipet and wash down the sides of the vial to ensure full recovery of the protein into solution. It is recommended to reconstitute the lyophilized product with sterile 5 mM Acetic acid to a concentration of 0.1 mg/ml, which can be further diluted into other aqueous solutions.
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service