Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6040

Sigma-Aldrich

TIMP-1 human

recombinant, expressed in Sf9 cells, ≥92% (SDS-PAGE)

Synonym(s):

TIMP, TIMP recombinant, TIMP-1 protein, Tissue Inhibitor of matrix metalloproteinases-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in Sf9 cells

assay

≥92% (SDS-PAGE)

form

liquid

mol wt

28.0 kDa

packaging

pkg of 5 μg

manufacturer/tradename

Croda International PLC

impurities

Endotoxin, tested

NCBI accession no.

shipped in

wet ice

storage temp.

−70°C

Gene Information

human ... TIMP1(7076)

General description

TIMP1 (tissue inhibitor of metalloproteinase 1) is a glycoprotein and is a member of TIMP family of endogenous MMP (matrix metalloproteinase) inhibitors. Humans contain four TIMPs. All TIMPs contain an N-terminal of 125 residues, a 65-residue C-terminal, and both these domains contain three disulfide bonds. The N-terminal domain inhibits MMPs by folding into a separate unit.[1] TIMP1 is encoded by the gene mapped to human chromosome Xp11.23.[2]

Biochem/physiol Actions

TIMP1 functions as a poor inhibitor of MT1 (membrane type 1)-MMP, MT3-MMP, MT5-MMP and MMP-19. It inhibits ADAM10 (a disintegrin and metalloproteinase domain-containing protein 10) protein. In human umbilical vein endothelial cells (HUVECs), the down-regulation of TIMP1 and up-regulation of MMP-3 results in aberrant endothelium-dependent vasodilation, EC (endothelial cell) death, and endothelial interruption in a FOXO3 (forkhead box O3)-mediated manner. In patients with CAD (coronary artery disease) and acute coronary syndrome (ACS), the urine levels of this protein are elevated.[1] This protein interacts with Bcl-2 (B cell lymphoma) protein, and induces apoptosis in lung adenocarcinoma cells.[3] The plasma levels of this protein are increased in patients with obesity and obesity-related disorders, such as steatosis, where it participates in pathogenesis of diet-induced hepatic steatosis and glucose intolerance.[4]

Physical form

In 50 mM Tris-HCl, pH 7; 200 mM NaCl; 5 mM CaCl?; 1 ?M ZnCl?; 0.05% Brij® L23; 0,05% NaN?

Legal Information

Brij is a registered trademark of Croda International PLC

hcodes

Hazard Classifications

Aquatic Chronic 3

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tissue Inhibitor Of Matrix Metalloproteinase-1 Is Required for High-Fat Diet-Induced Glucose Intolerance and Hepatic Steatosis in Mice.
Fj?re E, et al.
PLoS ONE, 10(7) (2015)
Polymorphic X-chromosome inactivation of the human TIMP1 gene.
Anderson CL and Brown CJ
American Journal of Human Genetics, 65(3), 699-708 (1999)
Matrix metalloproteinase inhibitors as investigative tools in the pathogenesis and management of vascular disease.
Benjamin MM1 and Khalil RA.
EXS, 103, 209-279 (2012)
TIMP-1 Inhibits Apoptosis in Lung Adenocarcinoma Cells via Interaction with Bcl-2.
Nalluri S, et al.
PLoS ONE, 10(9) (2015)
Gary J Litherland et al.
Arthritis & rheumatology (Hoboken, N.J.), 66(8), 2175-2187 (2014-04-24)
To assess the role of glycogen synthase kinase 3 (GSK-3) as a regulator of cartilage destruction in human tissue and a murine model of osteoarthritis (OA). Surgical destabilization of the medial meniscus (DMM) was performed to induce experimental murine OA

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service