Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6034

Sigma-Aldrich

LBP from mouse

recombinant, expressed in CHO cells, ≥95% (SDS-PAGE)

Synonym(s):

Lipopolysaccharide binding protein

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

biological source

mouse

recombinant

expressed in CHO cells

assay

≥95% (SDS-PAGE)

form

lyophilized

mol wt

64.0 kDa

packaging

pkg of 10 μg

impurities

Endotoxin, tested

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... LBP(218846)

General description

Natural LBP is a 58KD glycoprotein produced in liver. It binds at lipid A of LPS with high affinity (10-9M) and reduced the cellular LPS effects at CD14+ cells (IL1ß, IL6, TNF?). It acts as opsonin for GRAM negative cells, LPS, neutrophiles and granulocytes.

Physical form

Lyophilized from a concentrated protein solution (1 mg/ml) containing PBS, pH 7.2

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in 10 μl of ultra-pure distilled water. This solution can then be diluted into other aqueous buffers.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fang Chen et al.
ChemMedChem, 14(15), 1457-1465 (2019-07-11)
Antimicrobial peptides (AMPs) that are able to neutralize toxins are promising antibiotics. In this study we investigated the role of structurally conserved amino acids in reduced human defensin 5 (HD5RED ), which is an endogenous peptide with antibacterial action and the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service