Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6033

Sigma-Aldrich

LBP human

recombinant, expressed in CHO cells, 90-95% (SDS-PAGE)

Synonym(s):

Lipopolysaccharide-binding protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in CHO cells

assay

90-95% (SDS-PAGE)

form

lyophilized

mol wt

58.0 kDa

packaging

pkg of 10 μg

technique(s)

ELISA: suitable

impurities

Endotoxin, tested

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... LBP(3929)

General description

Natural lipopolysaccharide binding protein (LBP) is a 58kDa glycoprotein produced in liver. This is a His-tagged protein.[1] It is part of a family of lipid-binding proteins.[2] The gene encoding this protein is localized on human chromosome 20q11.23.[3][4]

Biochem/physiol Actions

Lipopolysaccharide binding protein (LBP) binds at lipid A of lipopolysaccharide (LPS) with high affinity (10-9M) and reduces the cellular LPS effects at CD14+ cells (IL1β, IL6, TNF-α). It acts as opsonin for GRAM negative cells, LPS, neutrophils and granulocytes. It transfers LPS to its receptor and this leads to an active inflammatory response. LBP also binds to LPS-coated erythrocytes. The protein has a role in innate immune responses against Gram-negative bacterial infections.[1][5][6][2][4]

Physical form

Lyophilized from a concentrated protein solution (0.25 mg/ml) containing PBS, pH 7.1

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in sterile distilled H?O or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0 mg/ml.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Serum LBP Is Associated with Insulin Resistance in Women with PCOS.
Zhu Q
PLoS ONE, 11(1), e0145337-e0145337 (2016)
Copy number polymorphisms are not a common feature of innate immune genes.
Linzmeier RM and Ganz T
Genomics, 88(1), 122-126 (2006)
Robert S. Jack
Inflammation & Allergy Drug Targets null
Low levels of microbial translocation marker LBP are associated with sustained viral response after anti-HCV treatment in HIV-1/HCV co-infected patients.
Nystrom J
PLoS ONE, 10(3), e0118643-e0118643 (2015)
Robert S. Jack
CD14 in the Inflammatory Response null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service