Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6026

Sigma-Aldrich

UbcH5a human

recombinant, expressed in E. coli, ≥95% (SDS-PAGE)

Synonym(s):

UbcH

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

assay

≥95% (SDS-PAGE)

form

liquid

mol wt

17.0 kDa

packaging

pkg of 10 μg

concentration

~1.0 mg/mL

impurities

Endotoxin, tested

NCBI accession no.

shipped in

dry ice

Storage temp.

−70°C

Gene Information

human ... UB2D1(7321)

General description

UbcH5a is highly homologous to the S. cerevisiae Ubc4/5 family of E2 enzymes demonstrated to be essential for degradation of regulatory and abnormal proteins. UbcH5a, together with UbcH5b and UbcH5c, are the most active class of E2 enzymes in cell extracts and are associated with regulation of a number of transcription factors. Specifically, UbcH5a stimulates the conjugation of ubiquitin to the tumor suppressor p532. Useful for in vitro ubiquitinylation reactions. Typical enzyme concentration to support conjugation in vitro is 100 nM to 1 μM depending upon conditions.

Physical form

Supplied at a concentration of 1.0 mg/ml in 20 mM Tris-HCl, pH 7.5, containing 0.5 mM DTT.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service