Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6022

Sigma-Aldrich

Progranulin human

recombinant, expressed in HEK 293 cells, ≥90% (SDS-PAGE), ≥90% (HPLC), FLAG-tagged

Synonym(s):

Acrogranin, Granulin-1, Paragranulin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

biological source

human

recombinant

expressed in HEK 293 cells

assay

≥90% (HPLC)
≥90% (SDS-PAGE)

form

liquid

mol wt

88.0 kDa

packaging

pkg of 10 μg

concentration

0.5 mg/mL

impurities

Endotoxin, tested

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... GRN(2896)

General description

Progranulin, also called proepithelin and PC cell-derived growth factor, is a single precursor protein of granulins which are a family of secreted, glycosylated peptides that are cleaved from a single precursor protein with 7.5 repeats of a highly conserved 12-cysteine granulin/epithelin motif. Granulins are a variety of active, 6.0 kDa peptides and named granulin A (epithelin 1), granulin B (epithelin 2), granulin C, etc. Both the peptides and intact progranulin protein regulate cell growth. However, different members of the granulin protein family may act as inhibitors, stimulators, or have dual actions on cell growth. Granulin family members are important in normal development, wound healing, and tumorigenesis. Progranulin Human Recombinant fused to FLAG at C-terminus produced in HEK is a single, glycosylated, polypeptide chain containing 593 amino acids and having a molecular mass of 88.0 kDa. The Progranulin is purified by standard chromatographic techniques.

Physical form

Sterile filtered clear solution in PBS

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lorenz Thurner et al.
Digestive diseases and sciences, 59(8), 1733-1742 (2014-03-05)
Recently, we identified neutralizing autoantibodies against progranulin (PGRN) in a wide spectrum of rheumatic diseases including cases with enteropathic spondylarthritis. PGRN is a secreted protein with strong anti-inflammatory effects, believed to be mediated by the direct inhibition of TNF receptors
Masato Kanazawa et al.
Brain : a journal of neurology, 138(Pt 7), 1932-1948 (2015-04-04)
In the central nervous system, progranulin, a glycoprotein growth factor, plays a crucial role in maintaining physiological functions, and progranulin gene mutations cause TAR DNA-binding protein-43-positive frontotemporal lobar degeneration. Although several studies have reported that progranulin plays a protective role
Chenhui Zhou et al.
Journal of neuroinflammation, 12, 200-200 (2015-11-04)
Subarachnoid hemorrhage (SAH) is a devastating neurological injury with high morbidity and mortality that is mainly caused by early brain injury (EBI). Progranulin (PGRN) is known to be involved in various biological functions, such as anti-inflammation and tissue repair. This
Yohei Ichimura et al.
Arthritis & rheumatology (Hoboken, N.J.), 67(12), 3245-3255 (2015-08-08)
Progranulin is a growth factor that is active in wound repair and is an antagonist of tumor necrosis factor (TNF) receptors, regulating fibroblast activation, angiogenesis, and inflammation. Because long-standing activation of gene programs related to wound healing is a hallmark
Michael J Smout et al.
PLoS pathogens, 11(10), e1005209-e1005209 (2015-10-21)
Infection with the human liver fluke Opisthorchis viverrini induces cancer of the bile ducts, cholangiocarcinoma (CCA). Injury from feeding activities of this parasite within the human biliary tree causes extensive lesions, wounds that undergo protracted cycles of healing, and re-injury

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service