Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP5369

Sigma-Aldrich

VLK, GST tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

PKDCC, SGK493

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$372.00

$372.00


Usually ships in 2 weeks.


Select a Size

Change View
20 μG
$372.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

$372.00


Usually ships in 2 weeks.

recombinant

expressed in baculovirus infected Sf9 cells

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~69 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PKDCC(91461)

General description

VLK or vertebrate lonesome kinase is a protein kinase which is first expressed in E-cadherin-positive anterior visceral endoderm and mesendoderm, and later its expression is confined to E-cadherin-negative mesenchyme. VLK regulates the rate of protein export from the Golgi and thereby plays an important role in the formation of functional stroma by mesenchymal cells. Targeted disruption of VLK leads to a defect in lung development and delayed ossification of endochondral bone. VLK deficient mice display neonatal lethality due to respiratory failure, with a suckling defect arising from a cleft palate. VLK is required for the appropriate timing of flat proliferative chondrocyte differentiation.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service