Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP5367

Sigma-Aldrich

PTK9, GST tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

A6, TWF1, Twinfilin 1

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$372.00

$372.00


Usually ships in 2 weeks.


Select a Size

Change View
20 μG
$372.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

$372.00


Usually ships in 2 weeks.

recombinant

expressed in baculovirus infected Sf9 cells

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~64 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... TWF1(5756)

General description

PTK9 is an actin monomer-binding protein conserved from yeast to mammals which has been localized to cortical G-actin-rich structures that may be regulated by the small GTPase RAC1. PTK9 has tyrosine kinase activity similar to that of recombinant FGFR2, with optimal activity over pH 6.5 to 7.4 and a preference for manganese over magnesium as a divalent cofactor. PTK9 plays a central role in regulation of actin dynamics and cell motility in lymphoma cell homeostasis in vivo and represent as a bona fide lymphoma drug target.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service