Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP5364

Sigma-Aldrich

ULK4, GST tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

DKFZp434E1822, FAM7C1, FLJ20574, REC01035

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$278.00

$278.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
20 μG
$278.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$278.00


Usually ships in 2 weeks.

Request a Bulk Order

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~97 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... ULK4(54986)

General description

ULK4, or Unc-51-like kinase 4, is a Ser/Thr protein kinase that is a member of the APG1/unc-51/ULK1 subfamily. ULK4 contains five HEAT repeats and a catalytic kinase domain. The ULK4 gene is conserved in many species including chimpanzee, dog, cow, mouse, rat, chicken, zebrafish, A.thaliana, and rice. ULK4 has been implicated as a gene involved in the development of hydrocephalus. This neurologic disorder in animal models is being used to elucidate factors responsible for the excessive accumulation of cerebrospinal fluid in hydrocephalic humans.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service