Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP5357

Sigma-Aldrich

TESK1, GST tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$372.00

$372.00


Usually ships in 2 weeks.


Select a Size

Change View
20 μG
$372.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

$372.00


Usually ships in 2 weeks.

recombinant

expressed in baculovirus infected Sf9 cells

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~106 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... TESK1(7016)

General description

TESK1 or testis-specific kinase 1 is a serine/threonine protein kinase that contains an N-terminal protein kinase domain and a C-terminal proline-rich domain which is most closely related to those of the LIM motif-containing protein kinases (LIMKs). TESK1 protein can phosphorylate myelin basic protein and histone in vitro and plays an important role at and after the meiotic phase of spermatogenesis. TESK1 is mainly expressed in testicular germ cells.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service