Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP5354

Sigma-Aldrich

MOS, GST tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

MGC119962, MGC119963, MSV

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$372.00

$372.00


Usually ships in 2 weeks.


Select a Size

Change View
20 μG
$372.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

$372.00


Usually ships in 2 weeks.

recombinant

expressed in baculovirus infected Sf9 cells

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~66 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... MOS(4342)

General description

MOS or v-mos Moloney murine sarcoma viral oncogene homolog is a serine/threonine kinase that activate the MAP kinase cascade through direct phosphorylation of the MAP kinase activator MEK. The early site-specific phosphorylation of CPEB , which was catalyzed by Eg2 is essential for the polyadenylation of c-mos mRNA, subsequent translation of c-mos, and oocyte maturation. The interaction with and phosphorylation of MYOD by MOS enhances muscle differentiation. MOS is an essential component of the cytostatic factor that acts almost exclusively in the second meiotic metaphase arrest.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service