Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP5343

Sigma-Aldrich

CyclinA2, GST tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

CCN1, CCNA, CCNA2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~78 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CCNA2(890)

General description

CyclinA2 belongs to the highly conserved cyclin family whose members are characterized by a dramatic periodicity in protein abundance through the cell cycle and function as regulators of CDKs. CyclinA2 is an essential component of all embryonic and somatic cell cycles in mammals. CyclinA2 is highly expressed in the testis and certain myeloid leukemia cells. Cyclin A also plays a major role in the control of DNA replication (3). CyclinA2 binds and activates CDC2 or CDK2 and thus promotes both cell cycle G1/S and G2/M transitions.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service