Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SRP5329

Sigma-Aldrich

NEK1, active, GST tagged from mouse

PRECISIO®, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

D8Ertd790e, MGC189817, kat

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO®

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

56-84 nmol/min·mg

mol wt

~85 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

mouse ... Nek1(18004)

General description

NEK1 or NIMA (never in mitosis gene a)-related kinase 1 is a serine/threonine kinase involved in cell cycle regulation and is found in a centrosomal complex with FEZ1, a neuronal protein that plays a role in axonal development. NEK1 is involved early in the DNA damage response pathway. NEK1 cycles through the nucleus via its nuclear localization and export signals. NEK1 protein participates in different signaling pathways to regulate diverse cellular processes and plays an important role in the kidney where it has opened a new avenue for studying cystogenesis and identifying possible modes of therapy.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service