Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SRP5322

Sigma-Aldrich

GRK3, active, GST tagged human

PRECISIO®, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

ADRBK2, BARK2

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$401.00

$401.00


Usually ships in 2 weeks.

Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
10 μG
$401.00

About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

$401.00


Usually ships in 2 weeks.

Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO®

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

4.8-7.2 nmol/min·mg

mol wt

~84 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... ADRBK2(157)

General description

GRK3 or beta-adrenergic receptor kinase 2 specifically phosphorylates the agonist-occupied form of the beta-adrenergic and related G protein-coupled receptors. GRK3 is a member of the receptor kinase family that broadly serves to regulate receptor function. GRK3 has 85% amino acid similarity with beta adrenergic receptor kinase 1, with the protein kinase catalytic domain having 95% similarity. GRK3 is highly expressed in lung, heart, and adipose tissue. Single nucleotide polymorphisms (SNPs) in the promoter region of GRK3 have been associated with bipolar disorder.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service