Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SRP5306

Sigma-Aldrich

PIK3C3 (Vps34), active, GST tagged human

PRECISIO®, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

MGC61518, VPS34, hVps34

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO®

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

78-216 nmol/min·mg

mol wt

~128 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PIK3C3(5289)

General description

PIK3C3 or phosphoinositide-3-kinase is a member of the PI 3-kinase family that mediates the active diversion of proteins from the secretory pathway to vacuoles. PIK3C3 are involved in both receptor-mediated signal transduction and intracellular trafficking. In yeast, the vps34 gene product is a PI 3-kinase. PIK3C3 has a substrate specificity and ion requirements that are distinct from the other known mammalian PI 3-kinases. PIK3C3 mediate catalysis on membranes and suppress futile adenosine triphosphatase cycles. PIK3C3 appears to alternate between a closed cytosolic form and an open form on the membrane.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service