Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SRP5297

Sigma-Aldrich

PIP5K1B, active, GST tagged human

PRECISIO®, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

MSS4, STM7

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$401.00

$401.00


Usually ships in 2 weeks.

Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
10 μG
$401.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$401.00


Usually ships in 2 weeks.

Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO®

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

160-253 nmol/min·mg

mol wt

~96 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PIP5K1B(8395)

General description

PIP5K1B (phosphatidylinositol-4-phosphate 5-kinase, type I, beta) is a member of the phosphatidylinositol-4-phosphate 5-kinase family. PIP5K1B gene contains 17 exons and spans more than 300 kb. The seventeenth exon was found by RT-PCR, and are derived from the 3-prime untranslated region of the PRKACG gene which is located on 9q13 approximately 3 kb downstream of the STM7.I 3-prime untranslated region.The overexpression of PIP5K1B in COS-7 cells induces an increase in short actin fibers and a decrease in actin stress fibers.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service