Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SRP5283

Sigma-Aldrich

ALK6 (BMPR1B), active, GST tagged human

PRECISIO®, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

ALK-6, ALK6, CDw293

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO®

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

16.2-26.5 nmol/min·mg

mol wt

~68 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... BMPR1B(658)

General description

ALK6 (also known as BMPR1B) is a member of the transmembrane serine/threonine kinase that is the member of the bone morphogenetic protein (BMP) receptor, which is closely related to the activin receptors, ACVR1 and ACVR2. ALK6 is mainly involved in the endochondral bone formation and embryogenesis. ALK6 expressed in normal and cancerous prostate tissues and used in the endocrine therapy that given to the prostate cancer patients. ALK6 receptor trafficking also play a significant role in FOP pathogenesis and used in human T-cell differentiation.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service