SIRT2 is a member of the sirtuin family of proteins which are homologs to the yeast Sir2 protein. Sirtuin family contain a sirtuin core domain and are grouped into four classes with SIRT2 being a member of class I. Inhibition of SIRT2 can lead to neuroprotection in cellular and invertebrate models of Huntington′s disease. Huntington′s disease is characterized by increased sterol synthesis in neuronal cells and this process is reversed by SIRT2 inhibition. SIRT2 can deacetylate lys40 of alpha-tubulin both in vitro and in vivo. Knockdown of SIRT2 via small interfering RNA results in tubulin hyperacetylation.
Physical form
Supplied in 50 mM sodium phosphate, pH 7.0, 300 mM NaCl, 150 mM imidazole, 0.1 mM PMSF, 0.2 mM DTT, 25% glycerol.
Preparation Note
After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.
Sirtuin 2 mutations in human cancers impair its function in genome maintenance.
Head PE
The Journal of Biological Chemistry (2017)
Questions
Reviews
★★★★★ No rating value