Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SRP5245

Sigma-Aldrich

MLK3 (1-488), active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

MAP3K11, MGC17114, MLK-3, MLK3, PTK1, SPRK

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$536.00

$536.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
10 μG
$536.00

About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

$536.00


Usually ships in 2 weeks.

Request a Bulk Order

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

8-11 nmol/min·mg

mol wt

~83 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... MAP3K11(4296)

General description

MLK3 or Mixed-Lineage Kinase 3 is a mitogen-activated protein kinase kinase kinase capable of preferentially activating MAPK8/JNK kinase and functions as a positive regulator of JNK signaling pathway. MLK3 can directly phosphorylate and activate IkappaB kinase alpha and beta, and is found to be involved in the transcription activity of NF-kappaB mediated by Rho family GTPases and CDC42. MLK3 is a signal-integrating kinase with conventional MAP3K catalytic activity and additional noncatalytic functions that contribute to RAF/ERK signalling. MLK3 is a component of the BRAF/RAF1 complex and is required for integrity of the complex and for activation of ERK by the complex.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deborah N Chadee et al.
Nature cell biology, 6(8), 770-776 (2004-07-20)
The ERK group of mitogen-activated protein kinases (MAPKs) is essential for cell proliferation stimulated by mitogens, oncogenic ras and raf (ref. 1). All MAPKs are activated by MAP3K/MEK/MAPK core pathways and the Raf proto-oncoproteins, especially B-Raf, are ERK-specific MAP3Ks (refs
Y L Ing et al.
Oncogene, 9(6), 1745-1750 (1994-06-01)
We have identified a novel protein kinase, designated MLK-3, from human thymus using RT-PCR and cDNA library screening. The deduced open reading frame, derived from sequencing a 3.5 kb MLK-3 cDNA, encodes a protein of 847 amino acids with several

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service