Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SRP5229

Sigma-Aldrich

PKN3 active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

RP11-545E17.1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

2.5-3.5 nmol/min·mg

mol wt

~130 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PKN3(29941)

General description

PKN3 or PRK3 is a serine/threonine protein kinase which can associate with RHOA in a GTP-dependent manner. PKN3 can also interact with GRAF and GRAF2 via the SH3 domains and phosphorylate these proteins. PKN3 is a mediator of malignant growth of human prostate cancer cells and functions downstream of activated PI3K. PKN3 is upregulated in human tumor cells but not in surrounding nontumorigenic tissues. Inducible knockdown of PKN3 expression inhibited metastasis in a mouse prostate tumor model.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Shibata et al.
Journal of biochemistry, 130(1), 23-31 (2001-07-04)
PKNbeta is a novel isoform of PKNalpha, which is one of the target protein kinases for the small GTPase Rho. By yeast two-hybrid screening of a human embryonic kidney 293 cell cDNA library with the PKNbeta linker region containing proline-rich
Frauke Leenders et al.
The EMBO journal, 23(16), 3303-3313 (2004-07-30)
Chronic activation of the phosphoinositide 3-kinase (PI3K)/PTEN signal transduction pathway contributes to metastatic cell growth, but up to now effectors mediating this response are poorly defined. By simulating chronic activation of PI3K signaling experimentally, combined with three-dimensional (3D) culture conditions

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service