Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP5226

Sigma-Aldrich

PAD4, GST tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

PAD, PADI4, PDI4, PDI5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~96 kDa

NCBI accession no.

application(s)

genomic analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PADI4(23569)

General description

Peptidyl arginine deiminase 4 (PAD4) is composed of 663 amino acids encoded by 2238 base pairs of PADI4 cDNA. This protein is expressed in peripheral blood CD3+ T cells, CD20+ B cells, CD15+ neutrophils and CD68+ monocytes. It is expressed in hematopoietic tissues, such as spleen, thymus, peripheral blood leucocytes, fetal liver and bone marrow. The gene encoding this protein is localized on human chromosome 1p36.[1][2]

Biochem/physiol Actions

Peptidyl arginine deiminases (PADIs) are responsible for the post-translational conversion of peptidylarginine to citrulline, in the presence of calcium ions. In individuals with rheumatoid arthritis (RA), this gene is expressed in hematological cells and synovial tissues, and variant in this gene is linked with susceptibility to RA. The expression of this protein is linked with DNA hypermethylation in acute promyelocytic leukemia (APL).[1][2][3]

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Analysis Note

This protein is not assayed for enzymatic activity.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Localization of peptidylarginine deiminase 4 (PADI4) and citrullinated protein in synovial tissue of rheumatoid arthritis.
Chang X, et.al
Rheumatology (Oxford, England), 44(1), 40-50 (2005)
A novel PAD4/SOX4/PU.1 signaling pathway is involved in the committed differentiation of acute promyelocytic leukemia cells into granulocytic cells.
Song G, et.al
Oncotarget, 7(3), 3144-3157 (2016)
Functional haplotypes of PADI4, encoding citrullinating enzyme peptidylarginine deiminase 4, are associated with rheumatoid arthritis.
Suzuki A, et.al
Nature Genetics, 34(4), 395-402 (2003)
Pingxin Li et al.
The Journal of experimental medicine, 207(9), 1853-1862 (2010-08-25)
Neutrophils trap and kill bacteria by forming highly decondensed chromatin structures, termed neutrophil extracellular traps (NETs). We previously reported that histone hypercitrullination catalyzed by peptidylarginine deiminase 4 (PAD4) correlates with chromatin decondensation during NET formation. However, the role of PAD4
K Nakashima et al.
The Journal of biological chemistry, 274(39), 27786-27792 (1999-09-17)
Three types of peptidylarginine deiminase (PAD), which converts a protein arginine residue to a citrulline residue, are widely distributed in animal tissues. Little is known about PAD of hemopoietic cells. We found that PAD activity in human myeloid leukemia HL-60

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service