Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP5211

Sigma-Aldrich

KAT7, active, GST tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

HBO1, HBOA, KAT7, MYST2

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~110 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... KAT7(11143)

General description

KAT7 or K (lysine) acetyltransferase 5 is a signaling protein that belongs to the MYST family of histone acetyl transferases (HATs) and was originally isolated as an HIV-1 TAT-interactive protein which play important roles in regulating chromatin remodeling, transcription and other nuclear processes by acetylating histone and non-histone proteins. KAT7 acts as a transcriptional inhibitor and inhibited AR-mediated transactivation of reporter constructs in CV-1 and PC-3 cells. KAT7 is part of a multisubunit complex that can acetylate histones H3 and H4.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Sharma et al.
The Journal of biological chemistry, 275(45), 35200-35208 (2000-08-10)
The androgen receptor (AR), a member of the nuclear receptor superfamily, plays a central role in male sexual differentiation and prostate cell proliferation. Results of treating prostate cancer by androgen ablation indicate that signals mediated through AR are critical for
M Iizuka et al.
The Journal of biological chemistry, 274(33), 23027-23034 (1999-08-07)
The origin recognition complex (ORC) is an initiator protein for DNA replication, but also effects transcriptional silencing in Saccharomyces cerevisiae and heterochromatin function in Drosophila. It is not known, however, whether any of these functions of ORC is conserved in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service