SRP5207
MO25 α, GST tagged human
recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Synonym(s):
CAB39, CGI-66, FLJ22682
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352200
NACRES:
NA.32
recombinant
expressed in baculovirus infected Sf9 cells
assay
≥70% (SDS-PAGE)
form
buffered aqueous glycerol solution
mol wt
~63 kDa
NCBI accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... CAB39(51719)
General description
MO25α (mouse protein 25 alpha) is a 40-kDa protein that interacts with the STE20-related adaptor-alpha (STRADα) pseudo kinase to form a regulatory complex capable of stimulating the activity of the LKB1 tumor suppressor protein kinase. LKB1 plays a critical role in cell proliferation, polarity and energy metabolism. LKB1 is mutated in the inherited Peutz-Jeghers cancer syndrome (PJS). MO25α binds directly to a conserved Trp-Glu-Phe sequence at the STRADα C terminus, and markedly enhances the binding of STRADα to LKB1 thereby increasing LKB1 catalytic activity.
Physical form
Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.
Preparation Note
after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles
Storage Class
10 - Combustible liquids
wgk_germany
WGK 1
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Jérôme Boudeau et al.
Journal of cell science, 117(Pt 26), 6365-6375 (2004-11-25)
Mutations in the LKB1 tumour suppressor threonine kinase cause the inherited Peutz-Jeghers cancer syndrome and are also observed in some sporadic cancers. Recent work indicates that LKB1 exerts effects on metabolism, polarity and proliferation by phosphorylating and activating protein kinases
Christine C Milburn et al.
Nature structural & molecular biology, 11(2), 193-200 (2004-01-20)
Mouse protein 25 alpha (MO25 alpha) is a 40-kDa protein that, together with the STE20-related adaptor-alpha (STRAD alpha) pseudo kinase, forms a regulatory complex capable of stimulating the activity of the LKB1 tumor suppressor protein kinase. The latter is mutated
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service