Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP5203

Sigma-Aldrich

LC20, GST tagged from mouse

recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

MGC3505, MLC2, MRLC1, MYL9, MYRL2

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$374.00

$374.00


Usually ships in 2 weeks.

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
50 μG
$374.00

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.32

$374.00


Usually ships in 2 weeks.

Notify Me

Get notified when this item is ready to ship via email.

biological source

mouse

recombinant

expressed in E. coli

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~45 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

mouse ... Myl9(98932)

General description

LC20 is the myosin light chain that may regulate muscle contraction by modulating the ATPase activity of myosin heads. LC20 protein binds calcium and is activated by myosin light chain kinase. Two transcript variants encoding different isoforms have been found for LC20 and the deduced 172-amino acid protein is highly conserved, with only 3 differences between the human and chicken proteins. Light chain phosphorylation causes the folded monomeric form of myosin to extend and assemble into filaments. This observation established the involvement of the LC20 in conformational transitions of smooth muscle myosin.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P D Wagner et al.
The Journal of biological chemistry, 261(17), 7778-7783 (1986-06-15)
Phosphorylation of the 20,000-Da light chains, LC20, of vertebrate smooth muscle myosins is thought to be the primary mechanism for regulating the actin-activated ATPase activities of these myosins and consequently smooth muscle contraction. While actin stimulates the MgATPase activities of
K M Trybus et al.
The Journal of biological chemistry, 263(31), 16485-16492 (1988-11-05)
Light chain phosphorylation causes the folded monomeric form of myosin to extend and assemble into filaments. This observation established the involvement of the 20-kDa regulatory light chain (LC20) in conformational transitions of smooth muscle myosin. To further assess the role

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service