Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP5186

Sigma-Aldrich

HO2, GST tagged human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

HMOX2

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$270.00

$270.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
50 μG
$270.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$270.00


Usually ships in 2 weeks.

Request a Bulk Order

recombinant

expressed in E. coli

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~62 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... HMOX2(3163)

General description

HO2, or heme oxygenase 2, belongs to the heme oxygenase family that are essential enzyme in heme catabolism that cleave heme to form biliverdin which is subsequently converted to bilirubin by biliverdin reductase. HO2 is part of the calcium-sensitive potassium channel complex and enhances channel activity in normoxia. Knockdown of HO2 expression reduces channel activity, an effect that is reversed by carbon monoxide. Inhibition of calcium-sensitive potassium channels by hypoxia is dependent on HO2 expression and is augmented by HO2 stimulation.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paul J Kemp
Biochemical and biophysical research communications, 338(1), 648-652 (2005-09-03)
The physiological response of the carotid body is critically dependent upon oxygen-sensing by potassium channels expressed in glomus cells. One such channel is the large conductance, voltage- and calcium-dependent potassium channel, BK(Ca). Although it is well known that a decrease
Sandile E J Williams et al.
Science (New York, N.Y.), 306(5704), 2093-2097 (2004-11-06)
Modulation of calcium-sensitive potassium (BK) channels by oxygen is important in several mammalian tissues, and in the carotid body it is crucial to respiratory control. However, the identity of the oxygen sensor remains unknown. We demonstrate that hemoxygenase-2 (HO-2) is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service