Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP5184

Sigma-Aldrich

GRB2, GST tagged human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

ASH, EGFRBP-GRB2, Grb3-3, MST084, MSTP084

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~49 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... GRB2(2885)

General description

GRB2 is an adaptor protein that is the homolog of the Sem5 gene of C. elegans and Drk gene of Drosophila. GRB2 protein contains one SH2 domain and two SH3 domains and plays a role in growth factor-mediated signal transduction. GRB2 forms a complex with activated EGFR and the RAS-specific guanine nucleotide exchange factor SOS1 thereby mediating the growth factor-induced activation of RAS. GRB2 has been implicated in growth factor regulation of the cytoskeleton and DNA synthesis. DAB2 also binds to GRB2 and competes with SOS for GRB2 binding. GRB2 interacts with the hepatitis C virus nonstructural 5A (NS5A) protein and inhibits the activation of ERK1 and ERK2 by EGF.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S L Tan et al.
Proceedings of the National Academy of Sciences of the United States of America, 96(10), 5533-5538 (1999-05-13)
Although hepatitis C virus (HCV) infection is an emerging global epidemic causing severe liver disorders, the molecular mechanisms of HCV pathogenesis remain elusive. The NS5A nonstructural protein of HCV contains several proline-rich sequences consistent with Src homology (SH) 3-binding sites
E J Lowenstein et al.
Cell, 70(3), 431-442 (1992-08-07)
A cDNA clone encoding a novel, widely expressed protein (called growth factor receptor-bound protein 2 or GRB2) containing one src homology 2 (SH2) domain and two SH3 domains was isolated. Immunoblotting experiments indicate that GRB2 associates with tyrosine-phosphorylated epidermal growth

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service