Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP5175

Sigma-Aldrich

CDKN3, GST tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

CDI1, CIP2, FLJ25787, KAP, KAP1, MGC70625

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$362.00

$362.00


Usually ships in 2 weeks.


Select a Size

Change View
20 μG
$362.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

$362.00


Usually ships in 2 weeks.

recombinant

expressed in baculovirus infected Sf9 cells

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~51 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CDKN3(1033)

General description

CDKN3 is a member of the dual specificity protein phosphatase family that acts as a cyclin-dependent kinase inhibitor. CDKN3 has been shown to interact with and dephosphorylate specifically the CDK2 kinase thereby preventing it′s activation. CDKN3 has been reported to be deleted, mutated, or overexpressed in several kinds of cancers. Increased expression of CDKN3 leads to increased levels of kinase-associated phosphatase activity that inhibits the G(1)/S transition of the cell cycle by dephosphorylating the cyclin-dependent kinases.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mihai D Niculescu et al.
Journal of neurochemistry, 89(5), 1252-1259 (2004-05-19)
Choline is an important methyl donor and a component of membrane phospholipids. In this study, we tested the hypothesis that choline availability can modulate cell proliferation and the methylation of genes that regulate cell cycling. In several other model systems
Chau-Ting Yeh et al.
Biochemical and biophysical research communications, 305(2), 311-314 (2003-05-15)
The cyclin-dependent kinase (Cdk)-associated protein phosphatase (KAP) is a human dual-specificity protein phosphatase that dephosphorylates Cdk2 on a conserved threonine residue, T160, in a cyclin dependent manner. Several aberrant KAP transcripts with characteristic deletion regions have been identified in hepatocellular

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service