Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP5152

Sigma-Aldrich

14-3-3 α/β, untagged human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

14-3-3 α/β, GW128, HS1, KCIP-1, YWHAB

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in E. coli

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~29 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... YWHAB(7529)

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 50mM NaCl, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

14-3-3beta binds to and negatively regulates the tuberous sclerosis complex 2 (TSC2) tumor suppressor gene product, tuberin.
Shumway SD
The Journal of Biological Chemistry, 278(4), 2089-2092 (2003)
The pro-inflammatory cytokine 14-3-3e is a ligand of CD13 in cartilage.
Nefla M
Journal of Cell Science, 128(17), 3250-3262 (2015)
14-3-3 family members act coordinately to regulate mitotic progression.
Dalal SN
Cell Cycle, 3(5), 672-677 (2004)
[Genome-wide association study of allergic diseases in Russians of Western Siberia].
Freidin MB
Molekuliarnaia Biologiia, 45(3), 464-472 (2011)
14-3-3 facilitates Ras-dependent Raf-1 activation in vitro and in vivo.
Roy S
Molecular and Cellular Biology, 18(7), 3947-3955 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service