Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP5134

Sigma-Aldrich

SMAD5, GST tagged human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

DKFZp781C1895, DKFZp781O1323, Dwfc, JV5-1, MADH5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in E. coli

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~82 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... SMAD5(4090)

General description

SMAD5 is a member of the SMAD family and mediates signaling by the transforming growth factor-beta (TGFβ) superfamily and related ligands. SMAD5 plays a critical role in the signaling pathway by which TGFβ inhibits the proliferation of human hematopoietic progenitor cells. SMAD5 is up-regulated in gastric epithelial cells during the infection of the pathogen Helicobacter pylori and it mediates apoptosis of gastric epithelial cells induced by H. pylori infection. In mature human B cells, bone morphogenetic protein 6 (BMP-6) inhibits cell growth and rapidly induces phosphorylation of SMADs 5 and 8.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tomokazu Nagasako et al.
The Journal of biological chemistry, 278(7), 4821-4825 (2002-12-11)
The gastric pathogen Helicobacter pylori activates epithelial cell signaling pathways, and its infection induces changes in the expression of several genes in infected human gastric tissues. Recent studies have indicated that the ability of H. pylori to regulate epithelial cell
C H Heldin et al.
Nature, 390(6659), 465-471 (1997-12-11)
The recent identification of the SMAD family of signal transducer proteins has unravelled the mechanisms by which transforming growth factor-beta (TGF-beta) signals from the cell membrane to the nucleus. Pathway-restricted SMADs are phosphorylated by specific cell-surface receptors that have serine/threonine

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service