Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP5123

Sigma-Aldrich

RAC3, His tagged human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$270.00

$270.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
50 μG
$270.00

About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

$270.00


Usually ships in 2 weeks.

Request a Bulk Order

biological source

human

recombinant

expressed in E. coli

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~24 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... RAC3(5881)

General description

RAC3 is a GTPase which belongs to the RHO family of small GTP-binding proteins that regulate a diverse array of cellular events, including the control of cell growth, cytoskeletal reorganization, and the activation of protein kinases. RAC3 is primarily expressed in brain and is proposed to play a specific function in neuronal cells. Depletion of RAC3 induces stronger cell adhesion and dramatically increases the outgrowth of neurite-like protrusions whereas overexpression of RAC3 results in a contractile round morphology. RAC3 can exert its function through negatively affecting integrin-mediated cell-matrix adhesions. RAC3 shares with RAC1 the ability to interfere with cadherin-mediated adhesion.

Physical form

Supplied in 50mM sodium phosphate, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Encarnación Lozano et al.
Journal of cell science, 121(Pt 7), 933-938 (2008-03-06)
E-cadherin cell-cell adhesion plays a major role in the maintenance of the morphology and function of epithelial tissues. Modulation of E-cadherin function is an important process in morphogenesis and tumour de-differentiation. We have previously shown that constitutively active Rac1 induces
Amra Hajdo-Milasinovic et al.
Journal of cell science, 122(Pt 12), 2127-2136 (2009-06-06)
Rac1 and Rac3 are highly homologous regulatory proteins that belong to the small GTPases of the Rho family. Previously, we showed that Rac3 induces cell rounding and prevents neuronal differentiation, in contrast to its close relative Rac1, which stimulates cell

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service