Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP5121

Sigma-Aldrich

RAC1, GST tagged human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

MGC111543, MIG5, TC-25, p21-Rac1

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~47 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... RAC1(5879)

General description

RAC1 is a member of the Rho family and is a GTPase that is part of the small GTP-binding protein superfamily. RAC1 is involved in a diverse array of cellular events, including the control of cell growth, cytoskeletal reorganization, and the activation of protein kinases. Role of RAC1 in colorectal cancer has been reported after analysis of the protein expression level and activities of this protein in matched sets of tumor and non-tumor tissues. Overexpression of RAC1 leads to increased tumour growth in xenografts of human colorectal tumour cells. RAC1 is also involved in the regulation of critical cellular functions including organization of actin cytoskeleton, transcription control and cell cycle.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Teresa Gómez del Pulgar et al.
The international journal of biochemistry & cell biology, 39(12), 2289-2302 (2007-09-04)
The small GTPase Rac1 is involved in the regulation of critical cellular functions, such as transcription control, cell cycle, and organization of actin cytoskeleton. Rac1 signalling modulates cancer progression since its overexpression leads to an increased tumour growth of xenografts
Yoji Takami et al.
Digestive diseases and sciences, 53(2), 467-473 (2007-06-29)
Rho family GTPases play a pivotal role in the regulation of numerous cellular functions associated with malignant transformation and metastasis. To evaluate the role of these GTPases in colorectal cancer, the protein expression levels and activities of these proteins in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service