Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP5104

Sigma-Aldrich

NFATC1, GST tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

NF-ATC, NFAT2, NFATc

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~125 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... NFATC1(4772)

General description

NFATC1 is a member of the NFAT family of proteins which are Ca2+/calcineurin-responsive transcription factors primarily recognized for their central roles in T lymphocyte activation and cardiac valve development. NFAT consists of at least two components: a preexisting cytosolic component that translocates to the nucleus upon T-cell receptor (TCR) stimulation, and an inducible nuclear component. Proteins belonging to the NFAT family of transcription factors play a central role in inducible gene transcription during immune response. The product of NFATC1 is an inducible nuclear component and functions as a major molecular target for the immunosuppressive drugs such as cyclosporin A.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bong Sook Jhun et al.
Biochemical and biophysical research communications, 351(4), 986-992 (2006-11-14)
AMP-activated protein kinase (AMPK) is a key regulator of energy homeostasis and its activation during T cell receptor stimulation has recently been reported. In this study, we examined the role of AMPK in interleukin (IL)-2 production in T cells. Inhibition
Malte Buchholz et al.
The EMBO journal, 25(15), 3714-3724 (2006-07-29)
The nuclear factor of activated T cell (NFAT) proteins are a family of Ca2+/calcineurin-responsive transcription factors primarily recognized for their central roles in T lymphocyte activation and cardiac valve development. We demonstrate that NFATc1 is commonly overexpressed in pancreatic carcinomas

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service