Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SRP5070

Sigma-Aldrich

PRKX, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

PKX1

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$401.00

$401.00


Usually ships in 2 weeks.

Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
10 μG
$401.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$401.00


Usually ships in 2 weeks.

Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

127-173 nmol/min·mg

mol wt

~73 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PRKX(5613)

General description

PRKX is a serine/threonine protein kinase that is closely related to the catalytic subunit of the cAMP-dependent protein kinase. PRKX regulates epithelial morphogenesis during mammalian kidney development and PRKX expression markedly activates migration of cultured renal epithelial cells in the presence of cAMP. Aberrant adult kidney expression of PRKX is found in autosomal dominant polycystic kidney disease. PRKX is involved in macrophage and granulocyte maturation. Abnormal recombination between PRKX and a related pseudogene on chromosome Y is a frequent cause of sex reversal disorder in XX males and XY females.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Klink et al.
Human molecular genetics, 4(5), 869-878 (1995-05-01)
We have isolated a gene, PKX1, by virtue of its position within the candidate region for chondrodysplasia punctata in Xp22.3. Although data from one patient render it unlikely that PKX1 is the CDPX gene, this gene shows several interesting features.
Xiaohong Li et al.
Proceedings of the National Academy of Sciences of the United States of America, 99(14), 9260-9265 (2002-06-26)
The human protein kinase X gene (PRKX) is a member of an ancient family of cAMP-dependent serine/threonine kinases here shown to be phylogenetically distinct from the classical PKA, PKB/Akt, PKC, SGK, and PKG gene families. Renal expression of the PRKX

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service