Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SRP5057

Sigma-Aldrich

PDE1C, active, GST tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

Hcam3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

378-512 nmol/min·mg

mol wt

~99 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PDE1C(5137)

General description

PDE1C is a member of the phosphodiesterases (PDE) family which catalyzes the hydrolysis of cyclic nucleotides cAMP and cGMP to the corresponding nucleoside 5-prime-monophosphates. PDE1C binds both cAMP and cGMP with high affinity and hydrolyzes both substrates with similar rates of catalysis. PDE1C is a calmodulin-dependent PDE and is stimulated by calcium-calmodulin complex. PDE1C is expressed at high levels in human cardiac myocytes with an intracellular distribution distinct from that of other phosphodiesterases. PDE1C levels decrease in all conditions that inhibited cell proliferation and PDE1C is a useful marker in studying the dynamics of proliferation and migration of various cells.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Loughney et al.
The Journal of biological chemistry, 271(2), 796-806 (1996-01-12)
cDNAs corresponding to two human calcium, calmodulin (CaM)-regulated 3',5'-cyclic nucleotide phosphodiesterases (PDEs) were isolated. One, Hcam1 (PDE1A3), corresponds to the bovine 61-kDa CaM PDE (PDE1A2). The second, Hcam3 (PDE1C), represents a novel phosphodiesterase gene. Hcam1 encodes a 535-amino acid protein
Fabrice Vandeput et al.
The Journal of biological chemistry, 282(45), 32749-32757 (2007-08-30)
Isoforms in the PDE1 family of cyclic nucleotide phosphodiesterases were recently found to comprise a significant portion of the cGMP-inhibited cAMP hydrolytic activity in human hearts. We examined the expression of PDE1 isoforms in human myocardium, characterized their catalytic activity

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service