Recombinant full-length human JNK3 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. Further details are available in the Product Information Sheet under DOCUMENTS or at this link: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/385/168/srp5043dat.pdf
SRP5043
JNK3, active, GST tagged human
PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Synonym(s):
FLJ12099, FLJ33785, JNK3A, MAPK10, MGC50974, PRKM10, p493F12, p54bSAPK
About This Item
recombinant
expressed in baculovirus infected Sf9 cells
product line
PRECISIO® Kinase
assay
≥70% (SDS-PAGE)
form
buffered aqueous glycerol solution
specific activity
30-40 nmol/min·mg
mol wt
~71 kDa
NCBI accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... MAPK10(5602)
General description
Physical form
Preparation Note
Legal Information
Storage Class
10 - Combustible liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
Buongiorno, volevo sapere la posizione del tag GST, se all'N o al C-terminale. Grazie
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service