Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SRP5037

Sigma-Aldrich

GSG2 (470-end), active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

15-21 nmol/min·mg

mol wt

~64 kDa

NCBI accession no.

shipped in

dry ice

Storage temp.

−70°C

Gene Information

human ... GSG2(83903)

General description

Haspin is a protein kinase that regulates chromosome and spindle function during mitosis and meiosis. Haspin expression is detected in fetal liver, skin, kidney, small intestine and in all proliferating cells. Haspin phosphorylates H3 thr3 (H3T3ph) in human cell lines and depletion of Haspin by RNA interference reveals that Haspin is required for H3 thr3 phosphorylation in mitotic cells. Phosphorylation of H3T3ph by Haspin is necessary for chromosomal passenger complex (CPC) accumulation at centromeres. H3T3ph then positions the CPC at centromeres to regulate selected targets of Aurora B during mitosis.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Dai et al.
Genes & development, 19(4), 472-488 (2005-02-01)
Post-translational modifications of conserved N-terminal tail residues in histones regulate many aspects of chromosome activity. Thr 3 of histone H3 is highly conserved, but the significance of its phosphorylation is unclear, and the identity of the corresponding kinase unknown. Immunostaining
Fangwei Wang et al.
Science (New York, N.Y.), 330(6001), 231-235 (2010-08-14)
Aurora B is a component of the chromosomal passenger complex (CPC) required for correct spindle-kinetochore attachments during chromosome segregation and for cytokinesis. The chromatin factors that recruit the CPC to centromeres are unknown, however. Here we show that phosphorylation of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service