SRP5032
GCK, active, GST tagged human
PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Synonym(s):
BL44, MAP4K2, RAB8IP
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
12352200
NACRES:
NA.32
recombinant
expressed in baculovirus infected Sf9 cells
product line
PRECISIO® Kinase
assay
≥70% (SDS-PAGE)
form
buffered aqueous glycerol solution
specific activity
246-334 nmol/min·mg
mol wt
~125 kDa
NCBI accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... MAP4K2(5871)
General description
GCK is a serine/threonine protein kinase that is activated by TNFα and interacts with the TNF receptor-associated factor 2 (TRAF2). GCK is upstream of MAP kinases and particularly activates the SAPK pathway. Although GCK is found in many tissues, its expression in lymphoid follicles is restricted to the cells of germinal centre, where it may participate in B-cell differentiation. A mouse protein Rab8ip that has high homology to GCK can interact with the vesicular transport protein Rab8. Based on its interaction with Rab8, it is postulated that Rab8ip/GCK may modulate secretion in response to stress stimuli.
Physical form
Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.
Preparation Note
after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles
Legal Information
PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class
10 - Combustible liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
C M Pombo et al.
Nature, 377(6551), 750-754 (1995-10-26)
Eukaryotic cells respond to different extracellular stimuli by recruiting homologous signalling pathways that use members of the MEKK, MEK and ERK families of protein kinases. The MEKK-->MEK-->ERK core pathways of Saccharomyces cerevisiae may themselves be regulated by members of the
M Ren et al.
Proceedings of the National Academy of Sciences of the United States of America, 93(10), 5151-5155 (1996-05-14)
Rab8 is a small GTP-binding protein that plays a role in vesicular transport from the trans-Golgi network to the basolateral plasma membrane in polarized epithelial cells (MDCK), and to the dendritic surface in hippocampal neurons. As is the case for
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service